1-Boc-4-(4-aminophenoxy)piperidine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-{[1-(tert-Butoxycarbonyl)piperidin-4-yl]oxy}aniline
, tert-Butyl 4-(4-aminophenoxy)piperidine-1-carboxylate
076237-250MG
076237-1G
076237-5G
102545-142EA
2061.54
USD
102545-142
102545-140
102545-138
1-Boc-4-(4-aminophenoxy)piperidine ≥95%
1-Boc-4-(4-aminophenoxy)piperidine
Hazard: Irritant
Formula:
C₁₆H₂₄N₂O₃ MW: 292.3734 g/mol |
MDL Number:
MFCD04115023 CAS Number: 138227-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...