3-(4-Methylpiperazin-1-ylmethyl)benzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-{3-[(4-Methylpiperazin-1-yl)methyl]phenyl}methanamine

076225-5G 076225-10G 076225-1G
102545-076EA 218.46 USD
102545-076 102545-078 102545-080
3-(4-Methylpiperazin-1-ylmethyl)benzylamine ≥95%
3-(4-Methylpiperazin-1-ylmethyl)benzylamine

Hazard: Irritant

Formula: C₁₃H₂₁N₃
CAS Number: 515162-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR