Cyanoacetamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Cyanoacetamide

076203-100G 076203-10G
102544-970EA 155.38 USD
102544-970 102544-968
Cyanoacetamide ≥95%
Cyanoacetamide

Hazard: Irritant

Formula: C₃H₄N₂O
MW: 84.08 g/mol
MDL Number: MFCD00008024
CAS Number: 107-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR