(S)-(+)-2-(4-Isobutylphenyl)propionic Acid ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(+)-Ibuprofen(S)-2-(4-Isobutylphenyl)propanoic acid(S)-(+)-4-Isobutyl-alpha-methylphenylacetic Acid

076197-100G
102544-946EA 150.77 USD
102544-946
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid ≥95%
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid

Hazard: Irritant

Formula: C₁₃H₁₈O₂
MW: 206.29 g/mol
MDL Number: MFCD00069289
CAS Number: 51146-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR