2,6-Dimethylpyrazine ≥95%

Supplier: Matrix Scientific

076184-10G 076184-100G
102544-878EA 230.77 USD
102544-878 102544-876
2,6-Dimethylpyrazine ≥95%
2,6-Dimethylpyrazine

Hazard: Irritant

Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00006148
CAS Number: 108-50-9
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR