1,3,5-Tribromobenzene ≥95%

Supplier: Matrix Scientific

076150-100G 076150-500G
102544-720EA 78.46 USD
102544-720 102552-020
1,3,5-Tribromobenzene ≥95%
1,3,5-Tribromobenzene

Hazard: Irritant

Formula: C₆H₃Br₃
MW: 314.8 g/mol
Boiling Pt: 271 °C
Melting Pt: 121…124 °C
MDL Number: MFCD00000080
CAS Number: 626-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR