Phthalonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: Benzene-1,2-dicarbonitrile1,2-Benzenedicarbonitrile1,2-Dicyanobenzene

076149-100G
102544-716EA 133.85 USD
102544-716
Phthalonitrile ≥95%
Phthalonitrile

Hazard: Irritant

Formula: C₈H₄N₂
MW: 128.13 g/mol
MDL Number: MFCD00001771
CAS Number: 91-15-6
UN: 3439
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR