(R)-(-)-Glycidyl butyrate ≥95%

Supplier: Matrix Scientific

Synonyms: (-)-[(2R)-Oxiran-2-yl]methyl butanoate(R)-Oxiran-2-ylmethyl butyrate

076133-100G
102544-642EA 465.75 USD
102544-642
(R)-(-)-Glycidyl butyrate ≥95%
(R)-(-)-Glycidyl butyrate

Hazard: Irritant

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00075120
CAS Number: 60456-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR