Methyl-2-amino-3-chlorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-3-chlorobenzoate

076127-10G 076127-100G
102544-618EA 92.31 USD
102544-618 102544-620
Methyl-2-amino-3-chlorobenzoate ≥95%
Methyl 2-amino-3-chlorobenzoate

Hazard: Irritant

Formula: C₈H₈ClNO₂
MW: 185.6100 g/mol
MDL Number: MFCD06797833
CAS Number: 77820-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR