Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide ≥95%

Supplier: Matrix Scientific

Synonyms: (Diphenylphosphoryl)(mesityl)methanone

076103-100G
102544-508EA 289.23 USD
102544-508
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide ≥95%
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide

Purity: 95+%
CAS: 75980-60-8
MDL Number: MFCD00192110
Molecular Formula: C22H21O2P
Molecular Weight: 348,39
Hazard: Irritant

Formula: C₂₂H₂₁O₂P
MW: 348.38 g/mol
MDL Number: MFCD00192110
CAS Number: 75980-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR