N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl ((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)-L-valinate hydrochloride
, N-(2'-Cyanobiphenyl-4-ylmethyl) L-valine methyl ester monohydrochloride
076100-1G
076100-5G
076100-10G
102544-492EA
440
USD
102544-492
102544-490
102544-488
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride ≥95%
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride
Purity: 95+%
CAS: 482577-59-3
MDL Number: MFCD08460094
Molecular Formula: C20H23ClN2O2
Molecular Weight: 358,87
Hazard: Irritant
Formula:
C₂₀H₂₂N₂O₂ MW: 358.87 g/mol |
MDL Number:
MFCD08460094 CAS Number: 482577-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...