1,2,3-Trifluoro-5-nitrobenzene ≥95%
Supplier: Matrix Scientific
Synonyms:
3,4,5-Trifluoronitrobenzene
, 1,2,3-Trifluoro-5-nitrobenzene
076096-25G
076096-100G
102544-470EA
320
USD
102544-470
102833-900
1,2,3-Trifluoro-5-nitrobenzene ≥95%
1,2,3-Trifluoro-5-nitrobenzene
Hazard: Irritant
Formula:
C₆H₂F₃NO₂ MW: 177.08 g/mol |
MDL Number:
MFCD00456803 CAS Number: 66684-58-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...