2,2'-Azobis(2-methylpropionamidine)dihydrochloride (AAPH) ≥95%

Supplier: Matrix Scientific

076094-100G
102544-460EA 80 USD
102544-460
2,2'-Azobis(2-methylpropionamidine)dihydrochloride (AAPH) ≥95%
2,2'-Azobis(2-methylpropionamidine)dihydrochloride (AAPH)

Hazard: Flammable Solid

Formula: C₈H₁₈N₆·2HCl
MW: 271.19 g/mol
Melting Pt: 175…177 °C
MDL Number: MFCD00142725
CAS Number: 2997-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR