Lumefantrine ≥95%
Supplier: Matrix Scientific
Synonyms:
(Z)-2-(Dibutylamino)-1-(2,7-dichloro-9-(4-chlorobenzylidene)-9H-fluoren-4-yl)ethanol
, Benflumetol
, (9Z)-2,7-Dichloro-9-[(4-chlorophenyl)methylene]-alpha-[(dibutylamino)methyl]-9H-fluoren-4-methanol
076072-1G
102544-354EA
27.69
USD
102544-354
Lumefantrine ≥95%
Lumefantrine
Purity: 95+%
CAS: 82186-77-4
MDL Number: MFCD05662268
Molecular Formula: C30H32Cl3NO
Molecular Weight: 528,95
Hazard: Irritant
Formula:
C₃₀H₃₂Cl₃NO MW: 528.94 g/mol |
MDL Number:
MFCD05662268 CAS Number: 82186-77-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...