Adapalene ≥95%
Supplier: Matrix Scientific
Synonyms:
6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic Acid
, 6-(3-(Adamantan-1-yl)-4-methoxyphenyl)-2-naphthoic acid
076052-5G
076052-1G
102544-260EA
1167.69
USD
102544-260
102544-258
Adapalene ≥95%
Adapalene
Hazard: Irritant
Formula:
C₂₈H₂₈O₃ MW: 412.53 g/mol |
MDL Number:
MFCD03106112 CAS Number: 106685-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...