2,4-Diaminopyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: Pyrimidine-2,4-diaminePyrimidine-2,4-diyltriamine

076027-1G
102544-164EA 29.23 USD
102544-164
2,4-Diaminopyrimidine ≥95%
2,4-Diaminopyrimidine

Hazard: Irritant

Formula: C₄H₆N₄
MW: 110.12 g/mol
MDL Number: MFCD00038023
CAS Number: 156-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR