4-Acetylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-pyridyl ketone

075991-100G
102543-986EA 213.85 USD
102543-986
4-Acetylpyridine ≥95%
4-Acetylpyridine

Hazard: Irritant

Formula: C₇H₇NO
MW: 121.14 g/mol
MDL Number: MFCD00006433
CAS Number: 1122-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR