4-Fluoro-2-iodoaniline ≥95%

Supplier: Matrix Scientific

075966-100G
102543-860EA 269.23 USD
102543-860
4-Fluoro-2-iodoaniline ≥95%
4-Fluoro-2-iodoaniline

Hazard: Irritant

Formula: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD00042584
CAS Number: 61272-76-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR