4-Bromo-2-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

075965-1G
102543-850EA 15.38 USD
102543-850
4-Bromo-2-methylbenzaldehyde ≥95%
4-Bromo-2-methylbenzaldehyde

Hazard: Irritant

Formula: C₈H₇BrO
MDL Number: MFCD07787171
CAS Number: 24078-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR