2-(2,4-Difluorophenyl)pyridine ≥95%

Supplier: Matrix Scientific

075961-5G
102543-828EA 92.31 USD
102543-828
2-(2,4-Difluorophenyl)pyridine ≥95%
2-(2,4-Difluorophenyl)pyridine

Hazard: Irritant

Formula: C₁₁H₇F₂N
MW: 191.18 g/mol
MDL Number: MFCD07787526
CAS Number: 391604-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR