(R)-(-)-Methyl-3-hydroxybutyrate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl (R)-3-hydroxybutyrate(R)-Methyl 3-hydroxybutanoate

075905-5G
102543-562EA 15.38 USD
102543-562
(R)-(-)-Methyl-3-hydroxybutyrate ≥95%
(R)-(-)-Methyl 3-hydroxybutyrate

Hazard: Irritant

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 56…58 °C (11 mmHg)
Density: 1.055
Flash Pt: 73 °C
MDL Number: MFCD00063289
CAS Number: 3976-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR