3-Chloro-4-nitrotoluene ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-4-methyl-1-nitrobenzene
075898-10G
075898-5G
075898-100G
102543-526EA
90.77
USD
102543-526
102543-528
102543-530
3-Chloro-4-nitrotoluene ≥95%
3-Chloro-4-nitrotoluene
Hazard: Irritant
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol |
MDL Number:
MFCD02683043 CAS Number: 38939-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...