2-Acetylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-pyridyl ketone1-(Pyridin-2-yl)ethanone

075890-100G
102543-490EA 72.31 USD
102543-490
2-Acetylpyridine ≥95%
2-Acetylpyridine

Hazard: Irritant

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 189…190 °C
Density: 1.082
MDL Number: MFCD00006303
CAS Number: 1122-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR