p-Toluenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Toluenesulfonyl chlorideToluene-4-sulfonyl chlorideToluene-p-sulfonyl chlorideToluene-4-sulphonyl chlorideTosyl chloride4-Toluenesulphonyl chlorideToluene-p-sulphonyl chloride

075888-100G 075888-500G
102543-480EA 101.54 USD
102543-480 102664-072
p-Toluenesulfonyl chloride ≥95%
p-Toluenesulphonyl chloride

Hazard: Corrosive/Irritant/Moisture Sensitive

Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 146 °C (15 mmHg)
Melting Pt: 67…70 °C
Density: 1.35
MDL Number: MFCD00007450
CAS Number: 98-59-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR