Quinoxaline ≥95%
Supplier: Matrix Scientific
075886-100G
075886-10G
102543-468EA
36.92
USD
102543-468
102543-470
Quinoxaline ≥95%
Quinoxaline
Hazard: Irritant
Formula:
C₈H₆N₂ MW: 130.15 g/mol |
MDL Number:
MFCD00006719 CAS Number: 91-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...