2,3-Dichloro-6-methylpyridine ≥95%
Supplier: Matrix Scientific
075883-5G
075883-1G
102543-456EA
136.92
USD
102543-456
102543-454
2,3-Dichloro-6-methylpyridine ≥95%
2,3-Dichloro-6-methylpyridine
Hazard: Irritant
Formula:
C₆H₅Cl₂N MW: 162.0166 g/mol |
MDL Number:
MFCD09839232 CAS Number: 54957-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...