Ethyl-3-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl piperidine-3-carboxylateEthyl nipecotate

075855-100G
102543-324EA 75.38 USD
102543-324
Ethyl-3-piperidinecarboxylate ≥95%
Ethyl 3-piperidinecarboxylate

Hazard: Irritant/Air Sensitive

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 102…104 °C (7 mmHg)
Melting Pt: 165…167 °C
Density: 1.012
Flash Pt: 90 °C
MDL Number: MFCD00005991
CAS Number: 5006-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR