2-Phenylpropionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (±)-2-Phenylpropionic acid(±)-Hydratropic acidDL-2-Phenylpropionic acid(±)-2-Phenylpropanoic acid

075835-100G
102543-238EA 110.77 USD
102543-238
2-Phenylpropionic acid ≥95%
(±)-Hydratropic acid

Hazard: Irritant/Stench

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 264…266 °C
Melting Pt: –5 °C
Density: 1.1
MDL Number: MFCD00002650
CAS Number: 492-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR