2,6-Diaminopyrazine ≥95%

Supplier: Matrix Scientific

075820-5G
102543-168EA 1560 USD
102543-168
2,6-Diaminopyrazine ≥95%
2,6-Diaminopyrazine

Hazard: Irritant

Formula: C₄H₆N₄
MDL Number: MFCD09907864
CAS Number: 41536-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR