MATR075807

075807-100G
102543-100EA 62.77 USD
102543-100
MATR075807
Purity: 95+%
CAS: 586-76-5
MDL Number: MFCD00002529
Molecular Formula: C7H5BrO2
Molecular Weight: 201,03
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR