Procaine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Diethylamino)ethyl 4-Aminobenzoate2-(Diethylamino)ethyl 4-aminobenzoate4-Aminobenzoic acid 2-(Diethylamino)ethyl ester

075800-100G
102543-068EA 149.23 USD
102543-068
Procaine ≥95%
Procaine

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂
MW: 236.31 g/mol
Melting Pt: 62 °C
MDL Number: MFCD00007893
CAS Number: 59-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR