2-Bromobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: o-Bromobenzaldehyde

075788-100G
102543-018EA 36.92 USD
102543-018
2-Bromobenzaldehyde ≥95%
2-Bromobenzaldehyde

Hazard: Irritant/Air Sensitive

Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 228…230 °C
Melting Pt: 18…21 °C
Density: 1.597
Flash Pt: 95 °C
MDL Number: MFCD00003300
CAS Number: 6630-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR