Methyl-4-fluorosalicylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-fluoro-2-hydroxybenzoate

075716-5G
102542-676EA 169.23 USD
102542-676
Methyl-4-fluorosalicylate ≥95%
Methyl 4-fluorosalicylate

Hazard: Irritant

Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 38…40 °C
MDL Number: MFCD02093965
CAS Number: 392-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR