4,6-Difluorosalicylaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
2,4-Difluoro-6-hydroxybenzaldehyde
075691-5G
075691-1G
102542-558EA
826.15
USD
102542-558
102542-556
4,6-Difluorosalicylaldehyde ≥95%
4,6-Difluorosalicylaldehyde
Hazard: Irritant
Formula:
C₇H₄F₂O₂ MW: 158.1023 g/mol |
MDL Number:
MFCD03701048 CAS Number: 136516-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...