4-Chloro-(3-methoxycarbonyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-3-(methoxycarbonyl)phenylboronic acid4-Chloro-3-(methoxycarbonyl)benzeneboronic acid

075683-5G
102542-512EA 223.08 USD
102542-512
4-Chloro-(3-methoxycarbonyl)phenylboronic acid ≥95%
4-Chloro-(3-methoxycarbonyl)phenylboronic acid

Hazard: Irritant

Formula: C₈H₈BClO₄
MW: 214.4107 g/mol
MDL Number: MFCD06801690
CAS Number: 874219-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR