3-Bromo-2-fluorophenylacetic acid ≥95%
Supplier: Matrix Scientific
075681-5G
075681-1G
102542-502EA
330.77
USD
102542-502
102542-500
3-Bromo-2-fluorophenylacetic acid ≥95%
3-Bromo-2-fluorophenylacetic acid
Hazard: Irritant
Formula:
C₈H₆BrFO₂ MW: 233.0300 g/mol |
MDL Number:
MFCD09864663 CAS Number: 1375964-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...