2,6-Difluorobenzonitrile ≥95%
Supplier: Matrix Scientific
075661-10G
075661-100G
102542-402EA
41.54
USD
102542-402
102542-400
2,6-Difluorobenzonitrile ≥95%
2,6-Difluorobenzonitrile
Hazard: Toxic
Formula:
C₇H₃F₂N MW: 139.1 g/mol Boiling Pt: 197…198 °C Melting Pt: 28…32 °C Density: 1.246 Flash Pt: 80 °C |
MDL Number:
MFCD00001778 CAS Number: 1897-52-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...