2-Naphthylamine ≥94%

Supplier: Matrix Scientific

075648-5G
102542-334EA 100.05 USD
102542-334
2-Naphthylamine ≥94%
2-Naphthylamine

Hazard: Irritant/Store in Freezer

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 306 °C
Melting Pt: 111…113 °C
Density: 1.061
MDL Number: MFCD00004112
CAS Number: 91-59-8
UN: 1650
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR