2-Hydroxy-m-toluic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Cresotic acid2-Hydroxy-3-methylbenzoic acid3-Methylsalicylic acid

075644-100G
102542-314EA 58.65 USD
102542-314
2-Hydroxy-m-toluic acid ≥95%
2-Hydroxy-m-toluic acid

Hazard: Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00002448
CAS Number: 83-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR