Methyl-4-aminobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-aminobenzoate

075623-100G
102542-206EA 73.85 USD
102542-206
Methyl-4-aminobenzoate ≥95%
Methyl 4-aminobenzoate

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00007891
CAS Number: 619-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR