O-Methylisourea hemisulfate ≥95%

Supplier: Matrix Scientific

Synonyms: O-Methylisouronium sulfateO-Methylisouronium sulphate

075614-100G
102542-162EA 118.45 USD
102542-162
O-Methylisourea hemisulfate ≥95%
O-Methylisourea hemisulfate

Hazard: Irritant

Formula: C₂H₆N₂O·0.5H₂SO₄
MDL Number: MFCD00040594
CAS Number: 52328-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR