o-Anisidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminoanisole2-Anisidine2-Methoxyaniline

075608-100G 075608-10G
102542-128EA 903.28 USD
102542-128 102542-130
o-Anisidine ≥95%
o-Anisidine

Hazard: Toxic/Light Sensitive

Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 224…225 °C
Melting Pt: 5…6 °C
Density: 1.095
Flash Pt: 98 °C
MDL Number: MFCD00007688
CAS Number: 90-04-0
UN: 2431
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR