1,3-Dimethylbarbituric acid ≥95%

Supplier: Matrix Scientific

075566-100G
102541-940EA 73.85 USD
102541-940
1,3-Dimethylbarbituric acid ≥95%
1,3-Dimethylbarbituric acid

Hazard: Irritant

Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
Melting Pt: 121…123 °C
MDL Number: MFCD00006675
CAS Number: 769-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR