1-Ethylimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: N-Ethyl-1H-imidazoleN-Ethylimidazole1-Ethyl-1H-imidazole

075551-100G
102541-868EA 69.23 USD
102541-868
1-Ethylimidazole ≥95%
1-Ethylimidazole

Hazard: Corrosive/Stored under Argon/Light Sensitive/Refrigerate

Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 104 °C (262 mmHg)
Density: 0.995
MDL Number: MFCD00055389
CAS Number: 7098-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR