N-Boc-D-valine ≥95%
Supplier: Matrix Scientific
Synonyms:
Boc-D-Val-OH
075546-10G
075546-100G
102541-846EA
613.85
USD
102541-846
102541-844
N-Boc-D-valine ≥95%
N-Boc-D-valine
Hazard: Irritant
Formula:
C₁₀H₁₉NO₄ MW: 217.3 g/mol |
MDL Number:
MFCD00038282 CAS Number: 22838-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...