D-allo-Threonine ≥95%
Supplier: Matrix Scientific
Synonyms:
(2R,3R)-2-Amino-3-hydroxybutanoic acid
075507-5G
075507-1G
075507-10G
102541-666EA
70.77
USD
102541-666
102541-668
102541-670
D-allo-Threonine ≥95%
D-allo-Threonine
Hazard: Irritant
Formula:
C₄H₉NO₃ MW: 119.12 g/mol Melting Pt: 268 °C (dec.) |
MDL Number:
MFCD00004526 CAS Number: 24830-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...