2,3,3-Trimethylindolenine ≥95%

Supplier: Matrix Scientific

075495-10G 075495-100G
102541-616EA 207.69 USD
102541-616 102541-614
2,3,3-Trimethylindolenine ≥95%
2,3,3-Trimethylindolenine

Hazard: Irritant

Formula: C₁₁H₁₃N
MW: 159.23 g/mol
MDL Number: MFCD00005724
CAS Number: 1640-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR