2,5-Dichlorobenzaldehyde ≥95%

Supplier: Matrix Scientific

075477-100G
102541-536EA 212.31 USD
102541-536
2,5-Dichlorobenzaldehyde ≥95%
2,5-Dichlorobenzaldehyde

Hazard: Irritant

Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
MDL Number: MFCD00156140
CAS Number: 6361-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR