2-Nitro-p-xylene ≥95%

Supplier: Matrix Scientific

Synonyms: Nitro-p-xylene1,4-Dimethyl-2-nitrobenzene2,5-Dimethylnitrobenzene

075471-100G
102541-510EA 75.38 USD
102541-510
2-Nitro-p-xylene ≥95%
2-Nitro-p-xylene

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00024284
CAS Number: 89-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR