ω-Bromo-2-nitroacetophenone ≥95%

Supplier: Matrix Scientific

075431-5G 075431-1G 075431-10G
102541-326EA 275.38 USD
102541-326 102541-322 102541-324
ω-Bromo-2-nitroacetophenone ≥95%
ω-Bromo-2-nitroacetophenone

Hazard: Irritant

Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
MDL Number: MFCD00010294
CAS Number: 6851-99-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR